Ethyl-6-(2-furyl)-3-methyl-4-oxo-4,5,6,7-tetrahydro-1H-indole-2-carboxylate
Supplier: Matrix Scientific
080634-500MG
080634-1G
080634-5G
102670-490EA
646.15
USD
102670-490
102670-492
102670-494
Ethyl-6-(2-furyl)-3-methyl-4-oxo-4,5,6,7-tetrahydro-1H-indole-2-carboxylate
Ethyl-6-(2-furyl)-3-methyl-4-oxo-4,5,6,7-tetrahydro-1H-indole-2-carboxylate
Hazard: Irritant
Formula:
C₁₆H₁₇NO₄ |
MDL Number:
MFCD06752730 CAS Number: 27463-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...