1-Azaspiro[4.4]nonane hydrochloride
Supplier: Matrix Scientific
080633-500MG
080633-1G
080633-5G
102670-484EA
553.85
USD
102670-484
102670-486
102670-488
1-Azaspiro[4.4]nonane hydrochloride
1-Azaspiro[4.4]nonane hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClN |
MDL Number:
MFCD22421990 CAS Number: 1417567-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...