4-Amino-7-methyl-8-phenylpyrazolo[5,1-c][1,2,4]triazine-3-carbohydrazide
Supplier: Matrix Scientific
080630-500MG
080630-1G
102670-472EA
258.46
USD
102670-472
102670-474
4-Amino-7-methyl-8-phenylpyrazolo[5,1-c][1,2,4]triazine-3-carbohydrazide
4-Amino-7-methyl-8-phenylpyrazolo[5,1-c][1,2,4]triazine-3-carbohydrazide
Hazard: Irritant
Formula:
C₁₃H₁₃N₇O MW: 283.29 g/mol |
MDL Number:
MFCD22374933 CAS Number: 1374509-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...