6-Phenylisoxazolo[5,4-e]pyrazolo[5,1-c][1,2,4]triazin-3-amine

Supplier: Matrix Scientific

080629-500MG 080629-1G
102670-468EA 387.69 USD
102670-468 102670-470
6-Phenylisoxazolo[5,4-e]pyrazolo[5,1-c][1,2,4]triazin-3-amine
6-Phenylisoxazolo[5,4-e]pyrazolo[5,1-c][1,2,4]triazin-3-amine

Hazard: Irritant

Formula: C₁₂H₈N₆O
MW: 252.24 g/mol
MDL Number: MFCD22374932
CAS Number: 1374509-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR