4-Amino-8-phenylpyrazolo[5,1-c][1,2,4]triazine-3-carbothioamide

Supplier: Matrix Scientific

080628-500MG 080628-1G
102670-464EA 258.46 USD
102670-464 102670-466
4-Amino-8-phenylpyrazolo[5,1-c][1,2,4]triazine-3-carbothioamide
4-Amino-8-phenylpyrazolo[5,1-c][1,2,4]triazine-3-carbothioamide

Hazard: Irritant

Formula: C₁₂H₁₀N₆S
MDL Number: MFCD22374931
CAS Number: 1374509-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR