10-[4-(Diethylamino)phenyl]-3,4,9,10-tetrahydro-benzo[b]cyclopenta[e][1,4]diazepin-1(2H)-one
Supplier: Matrix Scientific
080624-500MG
080624-1G
102670-448EA
193.85
USD
102670-448
102670-450
10-[4-(Diethylamino)phenyl]-3,4,9,10-tetrahydro-benzo[b]cyclopenta[e][1,4]diazepin-1(2H)-one
10-[4-(Diethylamino)phenyl]-3,4,9,10-tetrahydro-benzo[b]cyclopenta[e][1,4]diazepin-1(2H)-one
Hazard: Irritant
Formula:
C₂₂H₂₅N₃O |
MDL Number:
MFCD22374928 CAS Number: 1374509-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...