2-Amino-4-methyl-7,8-dihydroquinazolin-5(6H)-one
Supplier: Matrix Scientific
080621-500MG
080621-1G
080621-5G
102670-432EA
387.69
USD
102670-432
102670-434
102670-436
2-Amino-4-methyl-7,8-dihydroquinazolin-5(6H)-one
2-Amino-4-methyl-7,8-dihydroquinazolin-5(6H)-one
Hazard: Irritant
Formula:
C₉H₁₁N₃O |
MDL Number:
MFCD01130240 CAS Number: 1374509-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...