N'-(2-Cyano-5,5-dimethyl-3-oxocyclohex-1-en-1-yl)-N,N-dimethylimidoformamide

Supplier: Matrix Scientific

080619-500MG 080619-1G 080619-5G
102670-422EA 258.46 USD
102670-422 102670-424 102670-426
N'-(2-Cyano-5,5-dimethyl-3-oxocyclohex-1-en-1-yl)-N,N-dimethylimidoformamide
N'-(2-Cyano-5,5-dimethyl-3-oxocyclohex-1-en-1-yl)-N,N-dimethylimidoformamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O
MDL Number: MFCD01241848
CAS Number: 418809-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR