(2E)-2-[(Dimethylamino)methylene]cycloheptanone hydrochloride

Supplier: Matrix Scientific

080612-500MG 080612-1G 080612-5G
102670-392EA 387.69 USD
102670-392 102670-394 102670-396
(2E)-2-[(Dimethylamino)methylene]cycloheptanone hydrochloride
(2E)-2-[(Dimethylamino)methylene]cycloheptanone hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClNO
MDL Number: MFCD22421989
CAS Number: 1417368-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR