(2E)-2-[(Dimethylamino)methylene]cyclopentanone hydrochloride

Supplier: Matrix Scientific

080611-500MG 080611-1G 080611-5G
102670-386EA 387.69 USD
102670-386 102670-388 102670-390
(2E)-2-[(Dimethylamino)methylene]cyclopentanone hydrochloride
(2E)-2-[(Dimethylamino)methylene]cyclopentanone hydrochloride

Hazard: Irritant

Formula: C₈H₁₄ClNO
MDL Number: MFCD22421988
CAS Number: 1417574-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR