(2E)-2-[(Dimethylamino)methylene]-4-methylcyclohexanone hydrochloride
Supplier: Matrix Scientific
080610-500MG
080610-1G
080610-5G
102670-380EA
387.69
USD
102670-380
102670-382
102670-384
(2E)-2-[(Dimethylamino)methylene]-4-methylcyclohexanone hydrochloride
(2E)-2-[(Dimethylamino)methylene]-4-methylcyclohexanone hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₈ClNO |
MDL Number:
MFCD22421987 CAS Number: 1417366-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...