(2E)-2-[(Dimethylamino)methylene]cyclohexanone hydrochloride

Supplier: Matrix Scientific

080609-500MG 080609-1G 080609-5G
102670-374EA 387.69 USD
102670-374 102670-376 102670-378
(2E)-2-[(Dimethylamino)methylene]cyclohexanone hydrochloride
(2E)-2-[(Dimethylamino)methylene]cyclohexanone hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO
MDL Number: MFCD22421986
CAS Number: 1417574-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR