Ethyl-1-(4-amino-6-oxo-1,6-dihydropyrimidin-2-yl)piperidine-4-carboxylate

Supplier: Matrix Scientific

080607-500MG 080607-1G
102670-366EA 258.46 USD
102670-366 102670-368
Ethyl-1-(4-amino-6-oxo-1,6-dihydropyrimidin-2-yl)piperidine-4-carboxylate
Ethyl-1-(4-amino-6-oxo-1,6-dihydropyrimidin-2-yl)piperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₈N₄O₃
MDL Number: MFCD11083010
CAS Number: 1030520-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR