6-Amino-1-[3-(trifluoromethyl)phenyl]pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080605-500MG 080605-1G
102670-356EA 258.46 USD
102670-356 102670-358
6-Amino-1-[3-(trifluoromethyl)phenyl]pyrimidine-2,4(1H,3H)-dione
6-Amino-1-[3-(trifluoromethyl)phenyl]pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₁H₈F₃N₃O₂
MW: 271.2 g/mol
MDL Number: MFCD06809797
CAS Number: 19136-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR