6-Amino-1-(4-fluorophenyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080604-500MG 080604-1G
102670-352EA 258.46 USD
102670-352 102670-354
6-Amino-1-(4-fluorophenyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(4-fluorophenyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₀H₈FN₃O₂
MDL Number: MFCD03765647
CAS Number: 16357-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR