6-Amino-1-(3-morpholin-4-ylpropyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080602-500MG 080602-1G
102670-342EA 258.46 USD
102670-342 102670-344
6-Amino-1-(3-morpholin-4-ylpropyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(3-morpholin-4-ylpropyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₁H₁₈N₄O₃
MW: 254.29 g/mol
MDL Number: MFCD09439082
CAS Number: 924846-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR