6-Amino-1-(1,3-benzodioxol-5-ylmethyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080600-500MG 080600-1G 080600-5G
102670-330EA 258.46 USD
102670-330 102670-332 102670-334
6-Amino-1-(1,3-benzodioxol-5-ylmethyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(1,3-benzodioxol-5-ylmethyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₄
MW: 261.24 g/mol
MDL Number: MFCD06260108
CAS Number: 157927-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR