6-Amino-1-(4-methoxybenzyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080599-500MG 080599-1G
102670-326EA 193.85 USD
102670-326 102670-328
6-Amino-1-(4-methoxybenzyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(4-methoxybenzyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₃
MDL Number: MFCD06260112
CAS Number: 54052-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR