6-Amino-1-(2-furylmethyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

080598-500MG 080598-1G
102670-322EA 258.46 USD
102670-322 102670-324
6-Amino-1-(2-furylmethyl)pyrimidine-2,4(1H,3H)-dione
6-Amino-1-(2-furylmethyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₉H₉N₃O₃
MDL Number: MFCD02904574
CAS Number: 230644-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR