Ethyl-2-hydroxy-6-phenylnicotinate

Supplier: Matrix Scientific

080591-500MG 080591-1G
102670-298EA 215.38 USD
102670-298 102670-300
Ethyl-2-hydroxy-6-phenylnicotinate
Ethyl-2-hydroxy-6-phenylnicotinate

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MDL Number: MFCD22374916
CAS Number: 58787-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR