7-Amino-2-methyl[1,2,4]triazolo-[1,5-a]pyrimidin-5(4H)-one

Supplier: Matrix Scientific

080584-500MG 080584-1G
102670-274EA 301.54 USD
102670-274 102670-276
7-Amino-2-methyl[1,2,4]triazolo-[1,5-a]pyrimidin-5(4H)-one
7-Amino-2-methyl[1,2,4]triazolo-[1,5-a]pyrimidin-5(4H)-one

Hazard: Irritant

Formula: C₆H₇N₅O
MDL Number: MFCD06613003
CAS Number: 5899-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR