2-Chloro-N-[1-(2-furoyl)-2-methyl-1,2,3,4-tetra-hydroquinolin-4-yl]-N-phenylacetamide
Supplier: Matrix Scientific
080580-500MG
080580-1G
102670-260EA
193.85
USD
102670-260
102670-262
2-Chloro-N-[1-(2-furoyl)-2-methyl-1,2,3,4-tetra-hydroquinolin-4-yl]-N-phenylacetamide
2-Chloro-N-[1-(2-furoyl)-2-methyl-1,2,3,4-tetra-hydroquinolin-4-yl]-N-phenylacetamide
Hazard: Irritant
Formula:
C₂₃H₂₁ClN₂O₃ |
MDL Number:
MFCD22374912 CAS Number: 1374509-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...