2-[(4-Methylquinazolin-2-yl)amino]quinazolin-4-ol
Supplier: Matrix Scientific
080576-500MG
080576-1G
102670-244EA
161.54
USD
102670-244
102670-246
2-[(4-Methylquinazolin-2-yl)amino]quinazolin-4-ol
2-[(4-Methylquinazolin-2-yl)amino]quinazolin-4-ol
Hazard: Irritant
Formula:
C₁₇H₁₃N₅O |
MDL Number:
MFCD00808928 CAS Number: 263746-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...