4-Chloro-N-(2-methyl-1,2,3,4-tetrahydroquinolin-4-yl)-N-phenylbenzamide

Supplier: Matrix Scientific

080571-500MG 080571-1G
102670-218EA 161.54 USD
102670-218 102670-220
4-Chloro-N-(2-methyl-1,2,3,4-tetrahydroquinolin-4-yl)-N-phenylbenzamide
4-Chloro-N-(2-methyl-1,2,3,4-tetrahydroquinolin-4-yl)-N-phenylbenzamide

Hazard: Irritant

Formula: C₂₃H₂₁ClN₂O
MDL Number: MFCD01156106
CAS Number: 305864-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR