Sodium-4-methylpiperazine-1-carbodithioate

Supplier: Matrix Scientific

080570-500MG 080570-1G
102670-214EA 258.46 USD
102670-214 102670-216
Sodium-4-methylpiperazine-1-carbodithioate
Sodium-4-methylpiperazine-1-carbodithioate

Hazard: Irritant

Formula: C₆H₁₁N₂NaS₂
MW: 198.29 g/mol
MDL Number: MFCD01878293
CAS Number: 5712-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR