2-(4-Fluorobenzoyl)hydrazinecarbothioamide

Supplier: Matrix Scientific

080569-500MG 080569-1G
102670-210EA 193.85 USD
102670-210 102670-212
2-(4-Fluorobenzoyl)hydrazinecarbothioamide
2-(4-Fluorobenzoyl)hydrazinecarbothioamide

Hazard: Irritant

Formula: C₈H₈FN₃OS
MDL Number: MFCD01424279
CAS Number: 831-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR