4-(Bromomethyl)-1-(4-fluorobenzoyl)-2,2-dimethyl-1,2-dihydroquinoline

Supplier: Matrix Scientific

080568-500MG 080568-1G 080568-5G
102670-204EA 193.85 USD
102670-204 102670-206 102670-208
4-(Bromomethyl)-1-(4-fluorobenzoyl)-2,2-dimethyl-1,2-dihydroquinoline
4-(Bromomethyl)-1-(4-fluorobenzoyl)-2,2-dimethyl-1,2-dihydroquinoline

Hazard: Irritant

Formula: C₁₉H₁₇BrFNO
MDL Number: MFCD22374910
CAS Number: 1374509-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR