N-(6-Chloro-4-phenylquinazolin-2-yl)guanidine

Supplier: Matrix Scientific

080566-500MG 080566-1G 080566-5G
102670-192EA 193.85 USD
102670-192 102670-194 102670-196
N-(6-Chloro-4-phenylquinazolin-2-yl)guanidine
N-(6-Chloro-4-phenylquinazolin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₅H₁₂ClN₅
MDL Number: MFCD02816999
CAS Number: 92434-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR