N-(8-Methoxy-4-methylquinazolin-2-yl)guanidine
Supplier: Matrix Scientific
080565-500MG
080565-1G
080565-5G
102670-186EA
193.85
USD
102670-186
102670-188
102670-190
N-(8-Methoxy-4-methylquinazolin-2-yl)guanidine
N-(8-Methoxy-4-methylquinazolin-2-yl)guanidine
Hazard: Irritant
Formula:
C₁₁H₁₃N₅O |
MDL Number:
MFCD00980702 CAS Number: 329213-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...