N-(6-Ethoxy-4-methylquinazolin-2-yl)guanidine

080563-500MG 080563-1G 080563-5G
102670-174 102670-176 102670-178
N-(6-Ethoxy-4-methylquinazolin-2-yl)guanidine
N-(6-Ethoxy-4-methylquinazolin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₂H₁₅N₅O
MDL Number: MFCD00658000
CAS Number: 5361-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR