N-(6-Ethyl-4-methylquinazolin-2-yl)guanidine

Supplier: Matrix Scientific

080562-500MG 080562-1G 080562-5G
102670-168EA 193.85 USD
102670-168 102670-170 102670-172
N-(6-Ethyl-4-methylquinazolin-2-yl)guanidine
N-(6-Ethyl-4-methylquinazolin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₂H₁₅N₅
MDL Number: MFCD01564234
CAS Number: 5361-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR