N-(4,7-Dimethylquinazolin-2-yl)guanidine

Supplier: Matrix Scientific

080559-500MG 080559-1G 080559-5G
102670-152EA 258.46 USD
102670-152 102670-154 102670-156
N-(4,7-Dimethylquinazolin-2-yl)guanidine
N-(4,7-Dimethylquinazolin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₁H₁₃N₅
MDL Number: MFCD00521264
CAS Number: 37836-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR