2-(Methoxymethyl)-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

Supplier: Matrix Scientific

080557-500MG 080557-1G
102670-146EA 387.69 USD
102670-146 102670-148
2-(Methoxymethyl)-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol
2-(Methoxymethyl)-5-methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

Hazard: Irritant

Formula: C₈H₁₀N₄O₂
MDL Number: MFCD18067549
CAS Number: 105445-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR