Ethyl-2-(4-tert-butylbenzyl)-3-oxobutanoate

Supplier: Matrix Scientific

080546-500MG 080546-1G
102670-088EA 258.46 USD
102670-088 102670-090
Ethyl-2-(4-tert-butylbenzyl)-3-oxobutanoate
Ethyl-2-(4-tert-butylbenzyl)-3-oxobutanoate

Hazard: Irritant

Formula: C₁₇H₂₄O₃
MDL Number: MFCD16343936
CAS Number: 36600-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR