Ethyl-2-(2-methylbenzyl)-3-oxobutanoate

Supplier: Matrix Scientific

080545-500MG 080545-1G 080545-5G
102670-082EA 167.69 USD
102670-082 102670-084 102670-086
Ethyl-2-(2-methylbenzyl)-3-oxobutanoate
Ethyl-2-(2-methylbenzyl)-3-oxobutanoate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MW: 234.3 g/mol
Boiling Pt: 120…125 °C (0.05 Torr)
MDL Number: MFCD16317678
CAS Number: 104075-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR