Ethyl-2-(4-methylbenzyl)-3-oxobutanoate

Supplier: Matrix Scientific

080544-500MG 080544-1G 080544-5G
102670-076EA 167.69 USD
102670-076 102670-078 102670-080
Ethyl-2-(4-methylbenzyl)-3-oxobutanoate
Ethyl-2-(4-methylbenzyl)-3-oxobutanoate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MDL Number: MFCD13961900
CAS Number: 14305-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR