Ethyl-2-(4-isopropylbenzyl)-3-oxobutanoate
Supplier: Matrix Scientific
080543-500MG
080543-1G
080543-5G
102670-070EA
167.69
USD
102670-070
102670-072
102670-074
Ethyl-2-(4-isopropylbenzyl)-3-oxobutanoate
Ethyl-2-(4-isopropylbenzyl)-3-oxobutanoate
Hazard: Irritant
Formula:
C₁₆H₂₂O₃ Boiling Pt: 122°C/0.2 Torr Density: 1.015 |
MDL Number:
MFCD16334275 CAS Number: 93163-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...