Ethyl-4-cyano-2-(4-methoxybenzoyl)butanoate

Supplier: Matrix Scientific

080540-500MG 080540-1G 080540-5G
102670-052EA 193.85 USD
102670-052 102670-054 102670-056
Ethyl-4-cyano-2-(4-methoxybenzoyl)butanoate
Ethyl-4-cyano-2-(4-methoxybenzoyl)butanoate

Hazard: Irritant

Formula: C₁₅H₁₇NO₄
MDL Number: MFCD22374907
CAS Number: 35747-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR