Ethyl-2-benzoylpentanoate

Supplier: Matrix Scientific

080532-500MG 080532-1G 080532-5G
102670-008EA 169.23 USD
102670-008 102670-010 102670-012
Ethyl-2-benzoylpentanoate
Ethyl-2-benzoylpentanoate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MDL Number: MFCD19302967
CAS Number: 24317-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR