N-(3,4,5,6-Tetrahydro-2H-azepin-7-yl)ethanimidamide hydrochloride

Supplier: Matrix Scientific

080526-500MG 080526-1G
102669-978EA 387.69 USD
102669-978 102669-980
N-(3,4,5,6-Tetrahydro-2H-azepin-7-yl)ethanimidamide hydrochloride
N-(3,4,5,6-Tetrahydro-2H-azepin-7-yl)ethanimidamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClN₃
MDL Number: MFCD22421984
CAS Number: 1417357-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR