10-Methyl-1H-pyrrolo[2,1-c][1,4]benzodiazepine-3,5,11(2H,10H,11aH)-trione

Supplier: Matrix Scientific

080525-500MG 080525-1G
102669-974EA 258.46 USD
102669-974 102669-976
10-Methyl-1H-pyrrolo[2,1-c][1,4]benzodiazepine-3,5,11(2H,10H,11aH)-trione
10-Methyl-1H-pyrrolo[2,1-c][1,4]benzodiazepine-3,5,11(2H,10H,11aH)-trione

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₃
MDL Number: MFCD22374903
CAS Number: 128076-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR