6-Methyl-2-[(4,6,8-trimethylquinazolin-2-yl)amino]pyrimidin-4(3H)-one
Supplier: Matrix Scientific
080523-500MG
080523-1G
102669-968EA
193.85
USD
102669-968
102669-970
6-Methyl-2-[(4,6,8-trimethylquinazolin-2-yl)amino]pyrimidin-4(3H)-one
6-Methyl-2-[(4,6,8-trimethylquinazolin-2-yl)amino]pyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₆H₁₇N₅O |
MDL Number:
MFCD22374901 CAS Number: 330830-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...