N-(3,4-Dihydro-2H-pyrrol-5-yl)-β-alanine

Supplier: Matrix Scientific

080519-500MG 080519-1G
102669-958EA 516.92 USD
102669-958 102669-960
N-(3,4-Dihydro-2H-pyrrol-5-yl)-β-alanine
N-(3,4-Dihydro-2H-pyrrol-5-yl)-beta-alanine

Hazard: Irritant

Formula: C₇H₁₂N₂O₂
MW: 156.19 g/mol
MDL Number: MFCD15203776
CAS Number: 5249-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR