2-(4-tert-Butylphenyl)-3,4-dihydro-5H-1,3,4-benzotriazepin-5-one

Supplier: Matrix Scientific

080516-500MG 080516-1G
102669-948EA 258.46 USD
102669-948 102669-950
2-(4-tert-Butylphenyl)-3,4-dihydro-5H-1,3,4-benzotriazepin-5-one
2-(4-tert-Butylphenyl)-3,4-dihydro-5H-1,3,4-benzotriazepin-5-one

Hazard: Irritant

Formula: C₁₈H₁₉N₃O
MDL Number: MFCD22374898
CAS Number: 1374509-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR