1-(2-Hydroxyethyl)-2-methylpyrimido-[1,2-a]benzimidazol-4(1H)-one
Supplier: Matrix Scientific
080515-500MG
080515-1G
102669-944EA
258.46
USD
102669-944
102669-946
1-(2-Hydroxyethyl)-2-methylpyrimido-[1,2-a]benzimidazol-4(1H)-one
1-(2-Hydroxyethyl)-2-methylpyrimido-[1,2-a]benzimidazol-4(1H)-one
Hazard: Irritant
Formula:
C₁₃H₁₃N₃O₂ |
MDL Number:
MFCD09438503 CAS Number: 932232-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...