2-[(4-Methoxyphenyl)amino]-5,6-dimethylpyrimidin-4(3H)-one
Supplier: Matrix Scientific
080509-500MG
080509-1G
102669-924EA
193.85
USD
102669-924
102669-926
2-[(4-Methoxyphenyl)amino]-5,6-dimethylpyrimidin-4(3H)-one
2-[(4-Methoxyphenyl)amino]-5,6-dimethylpyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₃H₁₅N₃O₂ |
MDL Number:
MFCD22374897 CAS Number: 1240678-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...