5-Benzyl-6-methyl-2-[(4-methylphenyl)amino]pyrimidin-4(3H)-one
Supplier: Matrix Scientific
080508-500MG
080508-1G
102669-920EA
193.85
USD
102669-920
102669-922
5-Benzyl-6-methyl-2-[(4-methylphenyl)amino]pyrimidin-4(3H)-one
5-Benzyl-6-methyl-2-[(4-methylphenyl)amino]pyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₉H₁₉N₃O |
MDL Number:
MFCD16980794 CAS Number: 897305-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...