N-(3-Chloro-2-methylphenyl)-N'-(4,5-dimethyl-6-oxo-1,6-dihydropyrimidin-2-yl)guanidine

Supplier: Matrix Scientific

080506-500MG 080506-1G
102669-914EA 193.85 USD
102669-914 102669-916
N-(3-Chloro-2-methylphenyl)-N'-(4,5-dimethyl-6-oxo-1,6-dihydropyrimidin-2-yl)guanidine
N-(3-Chloro-2-methylphenyl)-N'-(4,5-dimethyl-6-oxo-1,6-dihydropyrimidin-2-yl)guanidine

Hazard: Irritant

Formula: C₁₄H₁₆ClN₅O
MDL Number: MFCD22374895
CAS Number: 488796-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR