7-(4-Chlorobenzyl)-3-(4-methoxybenzyl)-8-methyl-1,2,3,4-tetrahydro-6H-pyrimido[1,2-a][1,3,5]triazin-6-one

Supplier: Matrix Scientific

080505-500MG 080505-1G
102669-910EA 193.85 USD
102669-910 102669-912
7-(4-Chlorobenzyl)-3-(4-methoxybenzyl)-8-methyl-1,2,3,4-tetrahydro-6H-pyrimido[1,2-a][1,3,5]triazin-6-one
7-(4-Chlorobenzyl)-3-(4-methoxybenzyl)-8-methyl-1,2,3,4-tetrahydro-6H-pyrimido[1,2-a][1,3,5]triazin-6-one

Hazard: Irritant

Formula: C₂₂H₂₃ClN₄O₂
MW: 410.91 g/mol
MDL Number: MFCD22374894
CAS Number: 1374509-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR