Methyl (4-oxo-2,3,4,5-tetrahydro-1,5-benzoxazepin-3-yl)acetate

Supplier: Matrix Scientific

080501-500MG 080501-1G 080501-5G
102669-896EA 338.46 USD
102669-896 102669-898 102669-900
Methyl (4-oxo-2,3,4,5-tetrahydro-1,5-benzoxazepin-3-yl)acetate
Methyl (4-oxo-2,3,4,5-tetrahydro-1,5-benzoxazepin-3-yl)acetate

Hazard: Irritant

Formula: C₁₂H₁₃NO₄
MDL Number: MFCD13817366
CAS Number: 1120258-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR